Learn More
SAI Infusion Technologies Rat Femoral Vein Catheter - 76cm, for Rats 150g+
SDP
Catalog No. NC1686032
Encompass
Femoral Vein Catheter for Rats 150g or larger. 76cm PU catheter with a 4cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access witn a 23G blunt needle. Product is for laboratory animal research only.
Catalog No. NC1686032 Supplier SAI Infusion Technologies Supplier No. RFV06
Femoral Vein Catheter for Rats 150g or larger. 76cm PU catheter with a 4cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access witn a 23G blunt needle. Product is for laboratory animal research only.