Learn More
Biomatik Corporation Rat Ferritin, Mitochondrial (FTMT) ELISA Kit
Supplier:  Biomatik Corporation EKU04076
Encompass_Preferred
Rat Ferritin, Mitochondrial (FTMT) ELISA Kit for detecting FTMT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZUG8. Gene ID: 291470. Target Synonyms: MtF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7044