Learn More
BIOHIPPO Rat FGa Microsample
SDP
Catalog No. 5000314550
Encompass
Rat FGa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGa in Plasma It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 57 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat FGa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat FGa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat FGa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14550 Supplier BIOHIPPO Supplier No. BHE15211236
Rat FGa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGa in Plasma It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 57 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat FGa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat FGa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat FGa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color