Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat FGF-BP ELISA Kit

Catalog No. ERA16RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ERA16RBX5 5 x 96 Tests
ERA16RB 96 Tests
ERA16RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ERA16RBX5 Supplier Invitrogen™ Supplier No. ERA16RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Rat FGF-BP quantitates rat FGF-BP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat FGF-BP.

FGFBP1 encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9QY10
Assay Range 0.41-100 ng/mL
Assay Sensitivity 0.41 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein;
Gene ID (Entrez) 64535
Gene Symbol FGFBP1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target FGFBP1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.