Learn More
Sino Biological Recombinant Rat FGF-21 Protein (His Tag), HPLC-verified
Catalog No. 502992724
Encompass_Preferred
A DNA sequence encoding the Rat FGF21 (NP_570108.1) (Met1-Ser208) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2724 Supplier Sino Biological Supplier No. 83944R08H100
A DNA sequence encoding the Rat FGF21 (NP_570108.1) (Met1-Ser208) was expressed with a polyhistidine tag at the C-terminus.