Learn More
Sino Biological Recombinant Anti-FGFR4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80093R124100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat FGFR4/CD334 (rR FGFR4/CD334; Catalog#80093-R08H; Q498D6; Met1-Asp367).
Catalog No. 50-241-2459