Learn More
Sino Biological Recombinant Rat FGL1 Protein (His Tag)
Supplier:  Sino Biological 8A6965R08B50
Encompass_Preferred
A DNA sequence encoding the rat FGL1 (Q5M8C6.1) (Met1-Val314) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-2749