Learn More
American Research Products Inc Rat Fibrinogen Degradation Pro
Catalog No. 5000326431
Encompass
Rat Fibrinogen Degradation Product FDP ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26431 Supplier American Research Products Inc Supplier No. CSBE07942R
Rat Fibrinogen Degradation Product FDP ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich