Learn More
Biomatik Corporation Rat Fibrinopeptide A (FPA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11635
Encompass_Preferred
Rat Fibrinopeptide A (FPA) Mini sample ELISA Kit for detecting FPA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 370.4-30,000pg/ml. Sensitivity: 129.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06399; P14480; P02680. Gene ID: 24366; 24367. Target Synonyms: FP-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4953