Learn More
Biomatik Corporation Rat Fibrinopeptide B (FPB) ELISA Kit
Supplier:  Biomatik Corporation EKU04116
Encompass_Preferred
Rat Fibrinopeptide B (FPB) ELISA Kit for detecting FPB in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.23-100ng/ml. Sensitivity: 0.47ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P14480. Gene ID: 24366. Target Synonyms: FP-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4853