Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 1, Acidic (FGF1) CLIA Kit
Supplier:  Biomatik Corporation EKU09632
Encompass_Preferred
Rat Fibroblast Growth Factor 1, Acidic (FGF1) CLIA Kit for detecting FGF1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.09pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P61149. Gene ID: 25317. Target Synonyms: ECGF; AFGF; ECGFA; ECGFB; FGF-Alpha; FGFA; HBGF1; ECGFB; HBGF1; Acidic Fibroblast Growth Factor; heparin-binding growth factor 1; Endothelial Cell Growth Factor, Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7374