Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 11 (FGF11) ELISA Kit
Supplier:  Biomatik Corporation EKU04132
Encompass_Preferred
Rat Fibroblast Growth Factor 11 (FGF11) ELISA Kit for detecting FGF11 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R5L8. Gene ID: 170632. Target Synonyms: FHF3; Fibroblast Growth Factor Homologous Factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5053