Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 13 (FGF13) ELISA Kit
Supplier:  Biomatik Corporation EKU04138
Encompass_Preferred
Rat Fibroblast Growth Factor 13 (FGF13) ELISA Kit for detecting FGF13 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ERW3. Gene ID: 84488. Target Synonyms: FHF2; Fibroblast Growth Factor Homologous Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5485