Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 10 (FGF10) ELISA Kit
Supplier:  Biomatik Corporation EKU04129
Encompass_Preferred
Rat Fibroblast Growth Factor 10 (FGF10) ELISA Kit for detecting FGF10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70492. Gene ID: 25443. Target Synonyms: Keratinocyte growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6122