Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 12 (FGF12) ELISA Kit
Supplier:  Biomatik Corporation EKU04135
Encompass_Preferred
Rat Fibroblast Growth Factor 12 (FGF12) ELISA Kit for detecting FGF12 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61150. Gene ID: 170630. Target Synonyms: FGF12B; FHF1; Fibroblast Growth Factor Homologous Factor 1; Myocyte-Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6939