Learn More
American Research Products Inc Rat fibroblast growth factor 2
Supplier:  American Research Products Inc CSBE12170R1
Encompass
Rat fibroblast growth factor 23 (FGF23) ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29964