Learn More
American Research Products Inc Rat fibroblast growth factor 2
Supplier:  American Research Products Inc CSBE12170R
Encompass
Rat fibroblast growth factor 23 (FGF23) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-30013