Learn More
American Research Products Inc Rat fibroblast growth factor 2
Catalog No. 5000327687
Encompass
Rat fibroblast growth factor 21 (FGF21) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27687 Supplier American Research Products Inc Supplier No. CSBEL008627RA
Rat fibroblast growth factor 21 (FGF21) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich