Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 3 (FGF3) ELISA Kit
Supplier:  Biomatik Corporation EKU04166
Encompass_Preferred
Rat Fibroblast Growth Factor 3 (FGF3) ELISA Kit for detecting FGF3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R5L9. Gene ID: 170633. Target Synonyms: HBGF3; INT2; Heparin-binding growth factor 3; Murine Mammary Tumor Virus Integration Site (v-int-2) Oncogene Homolog; INT-2 Proto-Oncogene Protein; Proto-oncogene Int-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5487