Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 6 (FGF6) CLIA Kit
Supplier:  Biomatik Corporation EKU09723
Encompass_Preferred
Rat Fibroblast Growth Factor 6 (FGF6) CLIA Kit for detecting FGF6 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.59pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q8R5L5. Gene ID: 170700. Target Synonyms: HBGF6; HST2; HST2 Oncogene; Heparin Secretory-Transforming Protein 2; Heparin-Binding Growth Factor 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6007