Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 7 (FGF7) ELISA Kit
Supplier:  Biomatik Corporation EKU08693
Encompass_Preferred
Rat Fibroblast Growth Factor 7 (FGF7) ELISA Kit for detecting FGF7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1000pg/ml. Sensitivity: 5.37pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q02195. Gene ID: 29348. Target Synonyms: KGF; HBGF7; Keratinocyte Growth Factor; Heparin-binding growth factor 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7359