Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 9 (FGF9) CLIA Kit
Supplier:  Biomatik Corporation EKU09728
Encompass_Preferred
Rat Fibroblast Growth Factor 9 (FGF9) CLIA Kit for detecting FGF9 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.65pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P36364. Gene ID: 25444. Target Synonyms: GAF; HBFG9; Glia-Activating Factor; Heparin-Binding Growth Factor 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8453