Learn More
Biomatik Corporation Rat Fibroblast Growth Factor Receptor 1 (FGFR1) ELISA Kit
Supplier:  Biomatik Corporation EKU04190
Encompass_Preferred
Rat Fibroblast Growth Factor Receptor 1 (FGFR1) ELISA Kit for detecting FGFR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04589. Gene ID: 79114. Target Synonyms: CD331; FLT2, KAL2; CEK; FLG; BFGFR, N-SAM,Basic Fibroblast Growth Factor Receptor 1; Fms-Related Tyrosine Kinase-2; Pfeiffer Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6633