Learn More
Biomatik Corporation Rat Fibromodulin (FMOD) ELISA Kit
Supplier:  Biomatik Corporation EKU04197
Encompass_Preferred
Rat Fibromodulin (FMOD) ELISA Kit for detecting FMOD in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50609. Gene ID: 64507. Target Synonyms: SLRR2E; FM; Collagen-binding 59 kDa protein; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4570