Learn More
ILEX LIFE SCIENCES LLC RAT FIBRONECTIN PROTEIN 1 MG
SDP
Supplier:  ILEX LIFE SCIENCES LLC A11311MG
Encompass_Preferred
Rat Fibronectin, Purified Native Protein (plasma), 1 mg. APPLICATION: Fibronectin is useful for the induction of cell attachment to a variety of surfaces such as plastic and glass tissue culture labware, petri dishes, coverslips, microcarrier beads, etc. Fibronectin is useful for growth and maintenance of cells in low serum conditions. In general, the reconstituted fibronectin should be diluted with sterile physiological saline or serum-free medium to a concentration of 10-50 ug/ml. MOLECULAR WEIGHT: 220 kDa. PURITY: >95%. FORMULATION: Lyophilized from a concentrated 1 mg/ml solution containing 20mM Tris Cl pH-7.5. STORAGE: 4*C. SYNONYMS: Cold insoluble globulin, FINC, LETS, MSF, FN. SUMMARY: Fibronectin takes part as a general cell adhesion molecule by anchoring cells to collagen or proteoglycan substrates. Fibronectin organizes cellular interaction with the ECM by binding to different components of the extracellular matrix and to membrane-bound Fibronectin receptors on cell surfaces.
Catalog No. 50-273-4032