Learn More
Biomatik Corporation Rat Fibulin 3 (FBLN3) ELISA Kit
Supplier:  Biomatik Corporation EKU11773
Encompass_Preferred
Rat Fibulin 3 (FBLN3) ELISA Kit for detecting FBLN3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35568. Target Synonyms: EFEMP1; DHRD; DRAD; FBNL; MLVT; MTLV; S1-5; EGF-Containing Fibulin-Like Extracellular Matrix Protein 1; Fibrillin-Like; Extracellular protein S1-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8085