Learn More
Biomatik Corporation Rat Ficolin 1 (FCN1) CLIA Kit
Supplier:  Biomatik Corporation EKU09789
Encompass_Preferred
Rat Ficolin 1 (FCN1) CLIA Kit for detecting FCN1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 68.6-50,000pg/ml. Sensitivity: 27.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9WTS8. Target Synonyms: FCNM; Collagen/fibrinogen domain-containing protein 1; Ficolin-alpha; Ficolin-A; M-ficolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8433