Learn More
Biomatik Corporation Rat Ficolin 1 (FCN1) ELISA Kit
Supplier:  Biomatik Corporation EKU04218
Encompass_Preferred
Rat Ficolin 1 (FCN1) ELISA Kit for detecting FCN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 4.7-300ng/ml. Sensitivity: 1.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTS8. Target Synonyms: FCNM; Collagen/fibrinogen domain-containing protein 1; Ficolin-alpha; Ficolin-A; M-ficolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5542