Learn More
Sino Biological Anti-VEGFR1/FLT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80101T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat VEGFR1 / FLT-1 (rR VEGFR1 / FLT-1; Catalog#80101-R08H; P53767; Met1-Glu758). VEGFR1 / FLT-1 specific IgG was purified by Rat VEGFR1 / FLT-1 affinity chromatography.
Catalog No. 50-255-2829