Learn More
American Research Products Inc Rat FMS like tyrosine kinase 3
Catalog No. 5000326487
Encompass
Rat FMS-like tyrosine kinase 3 ligand Flt-3L ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26487 Supplier American Research Products Inc Supplier No. CSBE07336R
Rat FMS-like tyrosine kinase 3 ligand Flt-3L ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich