Learn More
Innovative Research Inc Rat Follicle Stim Hormone Kit
Supplier:  Innovative Research Inc IRTFSHKT
Encompass_Preferred
Rat Follicle Stimulating Hormone ELISA Kit - This kit is intended for the quantitative determination of Rat Follicle Stimulating Hormone in biofluid samples, such as serum, plasma and other biological fluids. This is a competitive inhibition ELISA in strip well format with reagents for up to 96 tests . FSH present in samples reacts with an anti-Rat FSH monoclonal capture antibody or antigen coated and dried on a microtiter plate. Following this, FSH conjugated to biotin competes with the antigen in the sample to bind the capture antibody or antigen. Unbound conjugate and sample antigen is washed away and bound conjugate is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is inverse proportional to the concentration of active FSH in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Rat FSH. For RUO.
Catalog No. 50-274-4008