Learn More
Sino Biological Rat FOLR1 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 81073R02H50
Encompass_Preferred
A DNA sequence encoding the rat Folr1 (NP_598211.1) (Met1-Met231) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2927