Learn More
Biomatik Corporation Rat Free Fatty Acid Receptor 3 (FFAR3) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11128
Encompass_Preferred
Rat Free Fatty Acid Receptor 3 (FFAR3) Mini sample ELISA Kit for detecting FFAR3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2GV46. Gene ID: 365228. Target Synonyms: FFA3R; GPR41; G Protein-Coupled Receptor 41. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8212