Learn More
American Research Products Inc Rat Free Testosterone F TESTO
Catalog No. 5000329185
Encompass
Rat Free Testosterone F-TESTO ELISA Kit from CUSABIO Detection Range 0 3 pg/mL-60 pg/mL Sensitivity 0 15 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29185 Supplier American Research Products Inc Supplier No. CSBE05097R
Rat Free Testosterone F-TESTO ELISA Kit from CUSABIO Detection Range 0 3 pg/mL-60 pg/mL Sensitivity 0 15 pg/mL Sample serum plasma Method Competitive