Learn More
Biomatik Corporation Rat Free Thyroxine, Ft4 Elisa Kit
Supplier:  Biomatik Corporation EKC31489
Encompass_Preferred
Rat Free Thyroxine, Ft4 Elisa Kit for detecting Free Thyroxine, Ft4 in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 4-64pmol/l. Sensitivity: 1.08pmol/l. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4348