Learn More
American Research Products Inc Rat Free Tri iodothyronine Fr
Catalog No. 5000329092
Encompass
Rat Free Tri-iodothyronine Free-T3 ELISA Kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma tissue homogenates cell lysates Method Competitive
Catalog No. 50-003-29092 Supplier American Research Products Inc Supplier No. CSBE05076R
Rat Free Tri-iodothyronine Free-T3 ELISA Kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma tissue homogenates cell lysates Method Competitive