Learn More
Sino Biological Anti-Follistatin-like 1/FSTL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80885T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat FSTL1 (rR FSTL1; Catalog#80885-R08H; NP_077345.1; Met1-Ile306). FSTL1 specific IgG was purified by Rat FSTL1 affinity chromatography.
Catalog No. 50-255-2756