Learn More
Biomatik Corporation Rat Fucosidase Alpha L1, Tissue (FUCa1) ELISA Kit
Supplier:  Biomatik Corporation EKU04281
Encompass_Preferred
Rat Fucosidase Alpha L1, Tissue (FUCa1) ELISA Kit for detecting FUCa1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17164. Gene ID: 24375. Target Synonyms: A-FU; aFU; Alpha-L-Fucosidase; Alpha-L-fucoside fucohydrolase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5551