Learn More
Biomatik Corporation Rat G Protein Coupled Receptor 131 (GPR131) ELISA Kit
Supplier:  Biomatik Corporation EKU10852
Encompass_Preferred
Rat G Protein Coupled Receptor 131 (GPR131) ELISA Kit for detecting GPR131 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.253ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q80T02. Gene ID: 338443. Target Synonyms: GPBAR1; GPCR; BG37; GPCR19; M-BAR; TGR5; G Protein-Coupled Bile Acid Receptor 1; G-protein coupled receptor GPCR19; Membrane-type receptor for bile acids. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7529