Learn More
BIOHIPPO Rat GAL9 Microsample
SDP
Catalog No. 5000314026
Encompass
Rat GAL9 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAL9 in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 096 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GAL9 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GAL9 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GAL9 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14026 Supplier BIOHIPPO Supplier No. BHE15212578
Rat GAL9 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAL9 in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 096 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GAL9 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GAL9 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GAL9 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color