Learn More
Biomatik Corporation Rat Galectin 1 (GAL1) ELISA Kit
Supplier:  Biomatik Corporation EKU04316
Encompass_Preferred
Rat Galectin 1 (GAL1) ELISA Kit for detecting GAL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.274ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11762. Gene ID: 56646. Target Synonyms: LGALS1; GBP; HLBP14; Galaptin; Lectin,Galactoside-Binding Soluble 1; 14 kDa laminin-binding protein; Lactose-binding lectin 1; Putative MAPK-activating protein PM12; S-Lac lectin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8966