Learn More
Biomatik Corporation Rat Galectin 7 (GAL7) ELISA Kit
Supplier:  Biomatik Corporation EKU10854
Encompass_Preferred
Rat Galectin 7 (GAL7) ELISA Kit for detecting GAL7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97590. Gene ID: 29518. Target Synonyms: LGALS7; PIG1; TP53I1; HKL-14; Lectin Galactoside-Binding Soluble 7; p53-induced gene 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5132