Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Gamma Globulin

Catalog No. PI31885 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31885 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31885 Supplier Invitrogen™ Supplier No. 31885
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Storage: After opening, dilute only enough antibody for a single day's use. Country of Origin: USA.

Rat IgG is a type of immunoglobulin G (IgG) antibody that is commonly used in scientific research and diagnostics. It is produced by rats in response to foreign antigens, and is widely used as a tool for detecting and measuring the presence of specific proteins or other molecules in biological samples.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Liquid
Product Type Rat Gamma Globulin
Quantity 10 mg
Concentration 11.3 mg/mL
Formulation PBS with no preservative; pH 7.6
For Use With (Application) Control, ChIP Assay, Immunoprecipitation, Flow Cytometry
Isotype Ig
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.