Learn More
American Research Products Inc Rat Gamma glutamyltranspeptida
Catalog No. 5000327825
Encompass
Rat Gamma-glutamyltranspeptidase 1 (GGT1) ELISA kit from CUSABIO Detection Range 4 7 mIU/mL-300 mIU/mL Sensitivity 1 2 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27825 Supplier American Research Products Inc Supplier No. CSBEL009394RA
Rat Gamma-glutamyltranspeptidase 1 (GGT1) ELISA kit from CUSABIO Detection Range 4 7 mIU/mL-300 mIU/mL Sensitivity 1 2 mIU/mL Sample serum plasma tissue homogenates Method Sandwich