Learn More
Biomatik Corporation Rat Gamma-Secretase Activating Protein (gSAP) ELISA Kit
Supplier:  Biomatik Corporation EKU04338
Encompass_Preferred
Rat Gamma-Secretase Activating Protein (gSAP) ELISA Kit for detecting gSAP in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZPQ9. Gene ID: 311984. Target Synonyms: PION; Pigeon Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8963