Learn More
BIOHIPPO Rat GAP43 Microsample
SDP
Catalog No. 5000315276
Encompass
Rat GAP43 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAP43 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GAP43 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GAP43 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GAP43 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15276 Supplier BIOHIPPO Supplier No. BHE15212732
Rat GAP43 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAP43 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GAP43 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GAP43 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GAP43 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color