Learn More
David Kopf Instruments RAT GAS ANESTHESIA HEAD HOLDER
SDP
Catalog No. NC1756717
Encompass
Model 929-B; Rat Gas Anesthesia Head Holder. Unit includes standard tooth bar. (Ear bars sold seperately). Option: Palate bar available upon request. Palate bar option is recommended for surgical procedures requiring clear access to the ear canal. *For rats 400 grams to 700 grams.
Catalog No. NC1756717 Supplier David Kopf Instruments Supplier No. 929B
Model 929-B; Rat Gas Anesthesia Head Holder. Unit includes standard tooth bar. (Ear bars sold seperately). Option: Palate bar available upon request. Palate bar option is recommended for surgical procedures requiring clear access to the ear canal. *For rats 400 grams to 700 grams.