Learn More
Biomatik Corporation Rat Gastrokine 1 (GKN1) ELISA Kit
Supplier:  Biomatik Corporation EKU04351
Encompass_Preferred
Rat Gastrokine 1 (GKN1) ELISA Kit for detecting GKN1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.069ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6SJV7. Gene ID: 297418. Target Synonyms: CA11; AMP18; BRICD1; FOV; Foveolin; 18 kDa antrum mucosa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4903