Learn More
BIOHIPPO Rat GDF3 Microsample
SDP
Catalog No. 5000315614
Encompass
Rat GDF3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GDF3 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 30 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GDF3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GDF3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GDF3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15614 Supplier BIOHIPPO Supplier No. BHE15211144
Rat GDF3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GDF3 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 30 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GDF3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GDF3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GDF3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color