Learn More
Raybiotech Inc Rat GDNF ELISA Kit
Supplier:  Raybiotech Inc ELRGDNF1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 60 pg/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects rat and human GDNF. Other species not determined.
Catalog No. 50-195-0247